3-Fluorosalicylaldehyde ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Fluoro-2-hydroxybenzaldehyde
L19324.03
L19324.06
L19324.03EA
155
EUR
L19324.03
L19324.06
3-Fluorosalicylaldehyde ≥98%
3-Fluorosalicylaldehyde
Purity: 98%
CAS No.: 394-50-3
Molecular Formula: C7H5FO2
Formula Weight: 140.12
Melting Point: 67-71°
Beilstein Reference: 1932806
Formula:
C₇H₅FO₂ MW: 140,12 g/mol Melting Pt: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003319 CAS Number: 394-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...