1-Bromo-4-hexyloxybenzene ≥97%

Supplier: Thermo Scientific

Synonyms: 1-(4-Bromophenoxy)hexane, 1-Bromo-4-(hexyloxy)benzene

B20085.03 B20085.06
B20085.03EA 0 EUR
B20085.03 B20085.06
1-Bromo-4-hexyloxybenzene ≥97%
1-Bromo-4-hexyloxybenzene

Purity: 97% CAS No.: 30752-19-3 Molecular Formula: C12H17BrO Formula Weight: 257.18 Flash Point: 110°(230°F) Refractive Index: 1.5240 MDL No.: MFCD00173753

Formula: C₁₂H₁₇BrO
MW: 257,18 g/mol
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00173753
CAS Number: 30752-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR