2,6-Dimethyl-4-nitroanisole ≥99%

Supplier: Thermo Fisher Scientific
Warning

H26204.03 H26204.06
H26204.03EA 35.1 EUR
H26204.03 H26204.06
2,6-Dimethyl-4-nitroanisole ≥99%
2,6-Dimethyl-4-nitroanisole

Purity: 99% CAS No.: 14804-39-8 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Melting Point: 85-92° MDL No.: MFCD00024537

Formula: C₉H₁₁NO₃
Melting Pt: 85…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00024537
CAS Number: 14804-39-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR