1-Dodecylboronic acid pinacol ester ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
2-Dodecyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H31186.03
H31186.06
H31186.03EA
0
EUR
H31186.03
H31186.06
1-Dodecylboronic acid pinacol ester ≥98%
1-Dodecylboronic acid pinacol ester
Purity: 98%
CAS No.: 177035-82-4
Molecular Formula: C18H37BO2
Formula Weight: 296.31
Refractive Index: 1.4410
Boiling Point: 123°/0.2mm
MDL No.: MFCD12546195
Formula:
C₁₈H₃₇BO₂ MW: 296.31 g/mol Boiling Pt: 123 °C (0.2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD12546195 CAS Number: 177035-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...