4-Methoxy-3-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H52660.MD H52660.03
H52660.MDEA 0 EUR
H52660.MD H52660.03
4-Methoxy-3-(trifluoromethyl)phenylboronic acid ≥98%
4-Methoxy-3-(trifluoromethyl)phenylboronic acid

Purity: 98% CAS No.: 149507-36-8 Molecular Formula: C8H8BF3O3 Formula Weight: 219.95 Melting Point: 198-202° MDL No.: MFCD07363790

Formula: C₈H₈BF₃O₃
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD07363790
CAS Number: 149507-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR