Isoquinolin-N-oxide ≥50% (w/v) in water

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Isoquinoline N-Oxide

L14794.06 L14794.14
L14794.06EA 0 EUR
L14794.06 L14794.14
Isoquinolin-N-oxide ≥50% (w/v) in water
Isoquinolin-N-oxide

CAS No.: 1532-72-5 Molecular Formula: C9H7NO Formula Weight: 145.16 Odour: Odourless Beilstein Reference: 114345 MDL No.: MFCD00006903

Formula: C₉H₇NO
MW: 145.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006903
CAS Number: 1532-72-5
EINECS: 216-242-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR