1-Methyl-2-indoleboronic acid 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(1-Methyl-1H-indol-2-yl)boronic acid
H53028.MD
H53028.03
H53028.MDEA
92.5
EUR
H53028.MD
H53028.03
1-Methyl-2-indoleboronic acid 95%
1-Methyl-2-indoleboronic acid
Purity: 95%
CAS No.: 191162-40-0
Molecular Formula: C9H10BNO2
Formula Weight: 174.99
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 108-112°
MDL No.: MFCD01114668
Formula:
C₉H₁₀BNO₂ MW: 174.99 g/mol Melting Pt: 108…112 °C |
MDL Number:
MFCD01114668 CAS Number: 191162-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...