Benzothiazol-2-ylmethanol ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole

H31631.03 H31631.06
H31631.03EA 44.5 EUR
H31631.03 H31631.06
Benzothiazol-2-ylmethanol ≥97%
Benzothiazol-2-ylmethanol

Purity: 97% CAS No.: 37859-42-0 Molecular Formula: C8H7NOS Formula Weight: 165.21 Melting Point: 96-100° MDL No.: MFCD00226293

Formula: C₈H₇NOS
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00226293
CAS Number: 37859-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR