4-Carboxyphenylboronic acid pinacol ester ≥97%
Supplier: Thermo Scientific
Synonyms:
4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
H26825.06
H26825.06EA
129
EUR
H26825.06
4-Carboxyphenylboronic acid pinacol ester ≥97%
4-Carboxyphenylboronic acid pinacol ester
Purity: 97%
CAS No.: 180516-87-4
Molecular Formula: C13H17BO4
Formula Weight: 248.08
Melting Point: 227-231°
MDL No.: MFCD01863710
Formula:
C₁₃H₁₇BO₄ MW: 248,08 g/mol Melting Pt: 227…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863710 CAS Number: 180516-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...