3-Chloro-N,N-dimethylaniline 95%
Supplier: Thermo Scientific
|
Danger
|
L08563.06
L08563.14
L08563.06EA
60.7
EUR
L08563.06
L08563.14
3-Chloro-N,N-dimethylaniline 95%
3-Chloro-N,N-dimethylaniline
Purity: 95%
CAS No.: 6848-13-1
Molecular Formula: C8H10ClN
Formula Weight: 155.63
Refractive Index: 1.5760
Boiling Point: 230-232°
UN No.: 2810
MDL No.: MFCD00018456
Formula:
C₈H₁₀ClN MW: 155,63 g/mol Boiling Pt: 230…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018456 CAS Number: 6848-13-1 EINECS: 229-935-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...