3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(3-(Dimethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-Dimethylcarbamoyl-4-fluorobenzeneboronic acid, 4-Fluoro-3-(dimethylcarbamoyl)phenylboronic acid
H52988.MD
H52988.03
H52988.MDEA
0
EUR
H52988.MD
H52988.03
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid ≥98%
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid
Purity: 98%
CAS No.: 874219-27-9
Molecular Formula: C9H11BFNO3
Formula Weight: 211.00
Melting Point: 146-148°
MDL No.: MFCD08235046
Formula:
C₉H₁₁BFNO₃ MW: 211,00 g/mol Melting Pt: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235046 CAS Number: 874219-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...