Allyl phenyl sulphide ≥97%
Supplier: Thermo Scientific
|
Warning
|
L04771.09
L04771.18
L04771.09EA
63.9
EUR
L04771.09
L04771.18
Allyl phenyl sulphide ≥97%
Allyl phenyl sulphide
Purity: 97%
CAS No.: 5296-64-0
Molecular Formula: C9H10S
Formula Weight: 150.24
Flash Point:
Formula:
C₉H₁₀S MW: 150,24 g/mol Boiling Pt: 223…225 °C Density: 1,024 Flash Pt: >98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014957 CAS Number: 5296-64-0 EINECS: 226-142-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...