3'-Bromo-5'-(trifluoromethyl)acetophenone 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethanone, 1-[3-bromo-5-(trifluoromethyl)phenyl]-, 1-[3-Bromo-5-(trifluoromethyl)phenyl]ethanone

H31854.03 H31854.06
H31854.03EA 140 EUR
H31854.03 H31854.06
3'-Bromo-5'-(trifluoromethyl)acetophenone 97%
3'-Bromo-5'-(trifluoromethyl)acetophenone

Purity: 97% Molecular Formula: C9H6BrF3O Formula Weight: 267.04 Refractive Index: 1.5045 MDL No.: MFCD11847339

Formula: C₉H₆BrF₃O
Melting Pt: <25 °C
Storage Temperature: Ambient
MDL Number: MFCD11847339
CAS Number: 154259-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR