2-Chloro-6-methylbenzylamine ≥97%
Supplier: Thermo Scientific
L11249.03
L11249.06
L11249.03EA
0
EUR
L11249.03
L11249.06
2-Chloro-6-methylbenzylamine ≥97%
2-Chloro-6-methylbenzylamine
Purity: 97%
CAS No.: 57264-46-7
Molecular Formula: C8H10ClN
Formula Weight: 155.63
Refractive Index: 1.5590
Boiling Point: 110-112°/15mm
UN No.: 2735
MDL No.: MFCD00052810
Formula:
C₈H₁₀ClN MW: 155,6300 g/mol |
MDL Number:
MFCD00052810 CAS Number: 57264-46-7 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...