3-Pyridinesulphonic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Pyridine-3-sulphonic acid
A13101.14
A13101.22
A13101.36
A13101.14EA
58.6
EUR
A13101.14
A13101.22
A13101.36
3-Pyridinesulphonic acid ≥98%
3-Pyridinesulphonic acid
Purity: 98+%
CAS No.: 636-73-7
Molecular Formula: C5H5NO3S
Formula Weight: 159.16
Melting Point: ca 350° dec.
Beilstein Reference: 120822
UN No.: 2585
MDL No.: MFCD00006381
Formula:
C₅H₅NO₃S MW: 159,16 g/mol Melting Pt: ∼ 350 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006381 CAS Number: 636-73-7 EINECS: 211-265-9 ADR: 8,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...