1-Nonanesulphonic acid sodium salt ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

L06407.06 L06407.14
L06407.06EA 63 EUR
L06407.06 L06407.14
1-Nonanesulphonic acid sodium salt ≥98%
1-Nonanesulphonic acid sodium salt

Purity: 98+% CAS No.: 35192-74-6 Molecular Formula: C9H19NaO3S Formula Weight: 230.30 Melting Point: 300° Beilstein Reference: 3738676

Formula: C₉H₁₉NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00025010
CAS Number: 35192-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR