2-Bromoethyl phenyl sulphide ≥97%

Warning

Synonyms: (2-Bromoethyl)(phenyl)sulphane

L11005.06 L11005.14
L11005.06EA 65.5 EUR
L11005.06 L11005.14
2-Bromoethyl phenyl sulphide ≥97%
2-Bromoethyl phenyl sulphide

Purity: 97% CAS No.: 4837-01-8 Molecular Formula: C8H9BrS Formula Weight: 217.12 Boiling Point: 140-141°/10mm Beilstein Reference: 2206188

Formula: C₈H₉BrS
MW: 217,12 g/mol
Boiling Pt: 140…141 °C (10 mmHg)
Density: 1,44
Storage Temperature: Ambient
MDL Number: MFCD00040826
CAS Number: 4837-01-8
EINECS: 225-422-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR