2-(Difluoromethoxy)benzylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(2-(Difluoromethoxy)phenyl)methanamine
L19165.03
L19165.MD
L19165.03EA
98.2
EUR
L19165.03
L19165.MD
2-(Difluoromethoxy)benzylamine 97%
2-(Difluoromethoxy)benzylamine
Purity: 97%
CAS No.: 243863-36-7
Molecular Formula: C8H9F2NO
Formula Weight: 173.16
Refractive Index: 1.4910
Formula:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236229 CAS Number: 243863-36-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...