4-(Difluoromethoxy)benzylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(4-(Difluoromethoxy)phenyl)methanamine
L19166.06
L19166.06EA
375
EUR
L19166.06
4-(Difluoromethoxy)benzylamine 97%
4-(Difluoromethoxy)benzylamine
Purity: 97%
CAS No.: 177842-14-7
Molecular Formula: C8H9F2NO
Formula Weight: 173.16
Refractive Index: 1.4900
Beilstein Reference: 4180270
UN No.: 2735
MDL No.: MFCD00236231
Formula:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236231 CAS Number: 177842-14-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...