3'-Acetamidoacetophenone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-Acetamidoacetophenone
A12188.14
A12188.22
A12188.14EA
59.8
EUR
A12188.14
A12188.22
3'-Acetamidoacetophenone ≥98%
3'-Acetamidoacetophenone
Purity: 98%
CAS No.: 7463-31-2
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Melting Point: 125-129°
Beilstein Reference: 2363443
Formula:
C₁₀H₁₁NO₂ MW: 177.20 g/mol Melting Pt: 125…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00032278 CAS Number: 7463-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...