4'-Acetamidoacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Acetamidoacetophenone, N-(4-Acetylphenyl)acetamide

L03430.09 L03430.18
L03430.09EA 33.5 EUR
L03430.09 L03430.18
4'-Acetamidoacetophenone ≥98%
4'-Acetamidoacetophenone

Purity: 98% CAS No.: 2719-21-3 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 166-172° Beilstein Reference: 639749

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
Melting Pt: 166…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00014965
CAS Number: 2719-21-3
EINECS: 220-320-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR