Dibenzyl carbonate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11464.09
A11464.18
A11464.30
A11464.09EA
99.5
EUR
A11464.09
A11464.18
A11464.30
Dibenzyl carbonate ≥98%
Dibenzyl carbonate
Purity: 98%
CAS No.: 3459-92-5
Molecular Formula: C15H14O3
Formula Weight: 242.27
Flash Point:
Formula:
C₁₅H₁₄O₃ Boiling Pt: 150…155 °C (0,8 mmHg) Melting Pt: 27…30 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014436 CAS Number: 3459-92-5 EINECS: 222-401-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...