4'-(1-Piperazinyl)acetophenone ≥94%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Piperazinoacetophenone, 1-(4-(Piperazin-1-yl)phenyl)ethanone
B22832.06
B22832.14
B22832.22
B22832.06EA
50.7
EUR
B22832.06
B22832.14
B22832.22
4'-(1-Piperazinyl)acetophenone ≥94%
4'-(1-Piperazinyl)acetophenone
Purity: 94%
CAS No.: 51639-48-6
Molecular Formula: C12H16N2O
Formula Weight: 204.27
Melting Point: 93-98°
UN No.: 3259
MDL No.: MFCD00005962
Formula:
C₁₂H₁₆N₂O MW: 204,27 g/mol Melting Pt: 93…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005962 CAS Number: 51639-48-6 EINECS: 257-332-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...