Dimolybdenum boride ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific

43680.22 43680.14
43680.22EA 350 EUR
43680.22 43680.14
Dimolybdenum boride ≥99.5% (metals basis)
Dimolybdenum boride
Formula: BMo₂
MDL Number: MFCD01310400
CAS Number: 12006-99-4
EINECS: 234-492-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR