1-Octadecyne 97
Supplier: Thermo Fisher Scientific
H33288.03
H33288.09
H33288.03EA
65.6
EUR
H33288.03
H33288.09
1-Octadecyne 97
1-Octadecyne
Formula:
C₁₈H₃₄ MW: 250.47 g/mol Boiling Pt: 313 °C Melting Pt: 21…24 °C Density: 0.803 Storage Temperature: Ambient |
MDL Number:
MFCD00015088 CAS Number: 629-89-0 EINECS: 211-114-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...