1-Decyl-3-methylimidazoliumtetrafluoroborate ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Decyl-3-methylimidazolium tetrafluoroborate

H59179.06 H59179.18
H59179.06EA 0 EUR
H59179.06 H59179.18
1-Decyl-3-methylimidazoliumtetrafluoroborate ≥97%
1-Decyl-3-methylimidazolium tetrafluoroborate
Formula: C₁₄H₂₇BF₄N₂
MW: 310.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038853
CAS Number: 244193-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR