2-Chloro-5-methylpyridine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-5-picoline

H59145.AA H59145.AC
H59145.AAEA 0 EUR
H59145.AA H59145.AC
2-Chloro-5-methylpyridine ≥97%
2-Chloro-5-methylpyridine
Formula: C₆H₆ClN
MW: 127,57 g/mol
Boiling Pt: 97 °C (30 mmHg)
Density: 1,169
Storage Temperature: Ambient
MDL Number: MFCD00792460
CAS Number: 18368-64-4
EINECS: 418-050-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR