2-Mercaptoethanesulphonic acid sodium salt (mesna) 96%

Supplier: Thermo Scientific
Warning

Synonyms: Sodium 2-mercaptoethanesulphonate

J63989.09 J63989.14
J63989.09EA 134 EUR
J63989.09 J63989.14
2-Mercaptoethanesulphonic acid sodium salt (mesna) 96%
2-Mercaptoethanesulphonic acid sodium salt (Mesna)

Purity: 96% CAS No.: 19767-45-4 Molecular Formula: C2H6NaO3S2 Formula Weight: 164.18 Physical Form: Powder Appearance: White Beilstein Reference: 3657828 Merck Reference: 14,5909 MDL No.: MFCD00007535


Water-soluble disulfide reducing agent. Cofactor in methyl transfer reactions

Formula: C₂H₅NaO₃S₂
MW: 164,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007535
CAS Number: 19767-45-4
EINECS: 243-285-9
Merck Index: 14,5909

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR