Pepstatin A
Supplier: Thermo Fisher Scientific
J60237.X0
J60237.MB
J60237.X0EA
74
EUR
J60237.X0
J60237.MB
Pepstatin A
Proteins and Peptides
Protease inhibitor that acts on pepsin, renin and cathepsin D.
- Oligopeptide
Purity: 98%
CAS No.: 26305-03-3
Molecular Formula: C₃₄H₆₃N₅₀₉
Formula Weight: 685.90
Storage Temperature: 2 to 8 °C
Physical Form: Powder
Melting Point: 233 °C.
Beilstein Reference: 2201362
Merck Reference: 14,7147
MDL No.: MFCD00060740
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...