N-(5-Bromo-6-methyl-2-pyridyl)thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
H34079.MD
H34079.03
H34079.MDEA
0
EUR
H34079.MD
H34079.03
N-(5-Bromo-6-methyl-2-pyridyl)thiourea ≥97%
N-(5-Bromo-6-methyl-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C7H8BrN3S
Formula Weight: 246.12
Melting Point: 238-242°
MDL No.: MFCD22125033
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...