N-(3,5-Dichloro-2-pyridyl)thiourea ≥97%
Supplier: Thermo Scientific
|
Danger
|
H34120.MD
H34120.03
H34120.MDEA
39.7
EUR
H34120.MD
H34120.03
N-(3,5-Dichloro-2-pyridyl)thiourea ≥97%
N-(3,5-Dichloro-2-pyridyl)thiourea
Purity: 97%
CAS No.: 31545-32-1
Molecular Formula: C6H5Cl2N3S
Formula Weight: 222.09
Melting Point: 164-168°
MDL No.: MFCD15525457
Formula:
C₆H₅Cl₂N₃S MW: 222,09 g/mol Melting Pt: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD15525457 CAS Number: 31545-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...