N-(4-Chloro-2-pyridyl)thiourea ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H33291.MD
H33291.03
H33291.MDEA
0
EUR
H33291.MD
H33291.03
N-(4-Chloro-2-pyridyl)thiourea ≥97%
N-(4-Chloro-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C6H6ClN3S
Formula Weight: 187.65
Melting Point: 182-187°
MDL No.: MFCD22380707
Formula:
C₆H₆ClN₃S MW: 187.65 g/mol Melting Pt: 182…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD22380707 CAS Number: 1824186-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...