1-(4-Cyanophenyl)-2-thiourea ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-(4-Cyanophenyl)thiourea
L12219.03
L12219.06
L12219.03EA
99.7
EUR
L12219.03
L12219.06
1-(4-Cyanophenyl)-2-thiourea ≥98%
1-(4-Cyanophenyl)-2-thiourea
Purity: 98%
CAS No.: 3460-55-7
Molecular Formula: C8H7N3S
Formula Weight: 177.23
Melting Point: ca 205° dec.
Beilstein Reference: 2718720
UN No.: 3439
MDL No.: MFCD00041175
Formula:
C₈H₇N₃S MW: 177,23 g/mol Melting Pt: ∼ 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041175 CAS Number: 3460-55-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...