Ethyl acetopyruvate ≥98%

Supplier: Thermo Scientific

Synonyms: Ethyl 2,4-dioxopentanoate, Ethyl 2,4-dioxovalerate

A13808.14 A13808.06 A13808.22
A13808.14EA 195 EUR
A13808.14 A13808.06 A13808.22
Ethyl acetopyruvate ≥98%
Ethyl acetopyruvate

Purity: 98% CAS No.: 615-79-2 Molecular Formula: C7H10O4 Formula Weight: 158.15 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4730 Melting Point: 16-18° Boiling Point: 108-109°/12mm Beilstein Reference: 607062 MDL No.: MFCD00009124

Formula: C₇H₁₀O₄
MW: 158,15 g/mol
Boiling Pt: 108…109 °C (12 mmHg)
Melting Pt: 16…18 °C
Density: 1,126
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00009124
CAS Number: 615-79-2
EINECS: 210-447-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR