N-Phenylurethane ≥98%
Supplier: Thermo Fisher Scientific
L05027.14
L05027.22
L05027.14EA
64.4
EUR
L05027.14
L05027.22
N-Phenylurethane ≥98%
N-Phenylurethane
Purity: 98%
CAS No.: 101-99-5
Molecular Formula: C6H5NHCO2CH2CH3
Formula Weight: 165.19
Refractive Index: 1.5375
Melting Point: 49-51°
Boiling Point: 237°
Beilstein Reference: 1942785
Merck Reference: 14,7320
MDL No.: MFCD00026806
Formula:
C₉H₁₁NO₂ Boiling Pt: 237 °C Melting Pt: 49…51 °C Density: 1,106 Storage Temperature: Ambient |
MDL Number:
MFCD00026806 CAS Number: 101-99-5 EINECS: 202-995-9 Merck Index: 14,7320 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...