1-Methyl-1-phenyl-2-thiourea ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: N-Methyl-N-phenylthiourea

B20824.09 B20824.88
B20824.09EA 0 EUR
B20824.09 B20824.88
1-Methyl-1-phenyl-2-thiourea ≥97%
1-Methyl-1-phenyl-2-thiourea

Purity: 97% CAS No.: 4104-75-0 Molecular Formula: C8H10N2S Formula Weight: 166.24 Odour: Odourless Melting Point: 100-104° UN No.: 2811 MDL No.: MFCD00086629

Formula: C₈H₁₀N₂S
MW: 166,24 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00086629
CAS Number: 4104-75-0
EINECS: 223-877-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR