N-(5-Iodo-2-pyridyl)thiourea ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H34009.MD
H34009.03
H34009.06
H34009.MDEA
0
EUR
H34009.MD
H34009.03
H34009.06
N-(5-Iodo-2-pyridyl)thiourea ≥97%
N-(5-Iodo-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C6H6IN3S
Formula Weight: 279.10
Melting Point: 219-225°
MDL No.: MFCD01315199
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...