Methyl vinyl sulphone 95% stabilised
Supplier: Thermo Scientific
|
Danger
|
Stabilisation:
Stabilised with 4-tert-butylphenol
L01519.03
L01519.06
L01519.03EA
40.1
EUR
L01519.03
L01519.06
Methyl vinyl sulphone 95% stabilised
Methyl vinyl sulphone
Stabilized with 200 ppm 4-tert-butylphenol
Purity: 95%
CAS No.: 3680-02-2
Molecular Formula: C3H6O2S
Formula Weight: 106.15
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₃H₆O₂S MW: 106,15 g/mol Boiling Pt: 115…117 °C (19 mmHg) Density: 1,212 Flash Pt: >109 °C (228 °F) |
MDL Number:
MFCD00007567 CAS Number: 3680-02-2 EINECS: 222-958-0 UN: 2810 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...