Carbamazepine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 298-46-4
Molecular Formula: C15H12N2O
Formula Weight: 236.27
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 189-192°
Merck Reference: 14,1781
MDL No.: MFCD00005073
Cytochrome P450 3A4 inducing anti-epileptic drug; ligand for the GABA-A receptor benzodiazepine modulatory site
Formula:
C₁₅H₁₂N₂O Melting Pt: 189…192 °C |
MDL Number:
MFCD00005073 CAS Number: 298-46-4 EINECS: 206-062-7 Merck Index: 14,1781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...