D-Glucoronic acid 98%
Supplier: Thermo Scientific
Synonyms:
D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
309880250
309880100
309881000
ACRO309880250EA
283
EUR
ACRO309880250
ACRO309880100
ACRO309881000
D-Glucoronic acid 98%
D-Glucoronic acid
Formula:
C₆H₁₀O₇ MW: 194,139 g/mol Melting Pt: 158…162 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00166981 CAS Number: 6556-12-3 EINECS: 229-486-4 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
Specific optical rotation | +35.5° to +37.5° (20°C, 589 nm) (c=6, H2O) (after 4 h) |
Titration with NaOH | ≥97.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...