D-Glucoronic acid ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
L14350.06
L14350.14
L14350.06EA
51.6
EUR
L14350.06
L14350.14
D-Glucoronic acid ≥98%
D-Glucoronic acid
Purity: 98+%
CAS No.: 6556-12-3
Molecular Formula: C6H10O7
Formula Weight: 194.14
Melting Point: ca 158° dec.
Beilstein Reference: 1727083
Merck Reference: 14,4465
Optical Rotation: +35° (c=6 in water)
MDL No.: MFCD00077778
Formula:
C₆H₁₀O₇ Melting Pt: ∼ 158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00166981 CAS Number: 6556-12-3 EINECS: 229-486-4 Merck Index: 14,4465 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...