Retinoic acid

Supplier: Cayman Chemical

Synonyms: all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin

11017-50 11017-500
CAYM11017-50EA 58.6 EUR
CAYM11017-50 CAYM11017-500
Retinoic acid
Retinoic acid
Formula: C₂₀H₂₈O₂
MW: 300.44 g/mol
MDL Number: MFCD00001551
CAS Number: 302-79-4
EINECS: 206-129-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR