4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

H52572.MD H52572.03
H52572.MDEA 0 EUR
H52572.MD H52572.03
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥97%
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine

Purity: 97% CAS No.: 151266-23-8 Molecular Formula: C5H4N5I Formula Weight: 261.03 MDL No.: MFCD03787931

Formula: C₅H₄N₅I
MW: 261,03 g/mol
MDL Number: MFCD03787931
CAS Number: 151266-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR