4-(Boc-amino)piperidine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

367620050 367620250
ACRO367620050EA 175 EUR
ACRO367620050 ACRO367620250
4-(Boc-amino)piperidine 98%
4-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00798171
CAS Number: 73874-95-0
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR