4-Amino-1-boc-piperidine-4-carboxylic acid 98%
Supplier: Thermo Scientific
Synonyms:
4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid
L19794.03
L19794.MD
L19794.03EA
204
EUR
L19794.03
L19794.MD
4-Amino-1-boc-piperidine-4-carboxylic acid 98%
4-Amino-1-Boc-piperidine-4-carboxylic acid
Purity: 98%
CAS No.: 183673-71-4
Molecular Formula: C11H20N2O4
Formula Weight: 244.29
Storage Temperature: 2°C to 8°C
Melting Point: ca 290° dec.
Beilstein Reference: 7588844
MDL No.: MFCD01318728
Formula:
C₁₁H₂₀N₂O₄ MW: 244,29 g/mol Melting Pt: ∼ 290 °C (dec.) |
MDL Number:
MFCD01318728 CAS Number: 183673-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...