4,5-Dicyano-2-(4-methylphenyl)imidazole ≥97%
Supplier: Thermo Scientific
|
Danger
|
H54298.MD
H54298.03
H54298.MDEA
0
EUR
H54298.MD
H54298.03
4,5-Dicyano-2-(4-methylphenyl)imidazole ≥97%
4,5-Dicyano-2-(4-methylphenyl)imidazole
Purity: 97%
Molecular Formula: C12H8N4
Formula Weight: 208.22
MDL No.: MFCD20265230
Formula:
C₁₂H₈N₄ MW: 208,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20265230 CAS Number: 63857-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...