4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester

H27292.MD H27292.03
H27292.MDEA 0 EUR
H27292.MD H27292.03
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester 95%
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester

Purity: 95% CAS No.: 1073354-22-9 Molecular Formula: C17H20BClN2O3 Formula Weight: 346.62 Melting Point: 99-101° MDL No.: MFCD06798267

Formula: C₁₇H₂₀BClN₂O₃
MW: 346,62 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD06798267
CAS Number: 1073354-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR