N,N-Dimethyl 4-boronobenzenesulphonamide 95%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(N,N-Dimethylsulphamoyl)phenylboronic acid, 4-(Dimethylsulphamoyl)benzeneboronic acid

H52936.MD H52936.03
H52936.MDEA 97.7 EUR
H52936.MD H52936.03
N,N-Dimethyl 4-boronobenzenesulphonamide 95%
N,N-Dimethyl 4-boronobenzenesulphonamide

Purity: 95% CAS No.: 486422-59-7 Molecular Formula: C8H12BNO4S Formula Weight: 229.06 Melting Point: 140-144° MDL No.: MFCD06659849

Formula: C₈H₁₂BNO₄S
MW: 229,06 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD06659849
CAS Number: 486422-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR