4-(Phenylazo)diphenylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Phenylazodiphenylamine
A18612.06
A18612.14
A18612.22
A18612.06EA
47.6
EUR
A18612.06
A18612.14
A18612.22
4-(Phenylazo)diphenylamine 97%
4-(Phenylazo)diphenylamine
Purity: 97%
CAS No.: 101-75-7
Molecular Formula: C18H15N3
Formula Weight: 273.34
Melting Point: 80-86°
MDL No.: MFCD00003023
Formula:
C₁₈H₁₅N₃ MW: 273,34 g/mol Melting Pt: 80…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003023 CAS Number: 101-75-7 EINECS: 202-972-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...