2-Aminoazotoluene
Supplier: Thermo Scientific
|
Danger
|
J60574.22
J60574.36
J60574.22EA
36.1
EUR
J60574.22
J60574.36
2-Aminoazotoluene
2-Aminoazotoluene
CAS No.: 97-56-3
Molecular Formula: C14H15N3
Formula Weight: 225.29
Physical Form: Powder
Appearance: Orange brown
Odour: Odourless
Beilstein Reference: 6506005
Merck Reference: 14,420
MDL No.: MFCD00007733
C.I. 11160
Formula:
C₁₄H₁₅N₃ MW: 225,29 g/mol Melting Pt: 101…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007733 CAS Number: 97-56-3 EINECS: 202-591-2 Merck Index: 14,420 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...